MAT113930000 Matrix Scientific

C-[1-(2-Methyl-piperidin-1-yl)-cyclopentyl]-methylamine

  • MDL number MFCD08688355
  • Cas Number : 946681-91-0
  • Molecular Weight : 196.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021663.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021663.html
Availability details for
Quantity
  • Availability
  • Add to Cart