MAT113930048 Matrix Scientific

N-[4-(Hydrazinocarbonyl)phenyl]-cyclopropanecarboxamide

  • MDL number MFCD03109882
  • Cas Number : 446278-50-8
  • Molecular Weight : 219.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023989.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023989.html
Availability details for
Quantity
  • Availability
  • Add to Cart