MAT113930070 Matrix Scientific

4-(4-Hydroxy-6-methyl-2-oxo-2H-pyridin-1-yl)-butyric acid

  • MDL number MFCD05256482
  • Cas Number : 685862-22-0
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021641.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021641.html
Availability details for
Quantity
  • Availability
  • Add to Cart