MAT113930117 Matrix Scientific

[(2-Methoxy-benzyl)-methyl-amino]-acetic acid

  • MDL number MFCD08688016
  • Cas Number : 55739-41-8
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021643.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021643.html
Availability details for
Quantity
  • Availability
  • Add to Cart