MAT113930156 Matrix Scientific

2-[(2-Naphthyloxy)methyl]piperidine

  • MDL number MFCD02663632
  • Cas Number : 383128-47-0
  • Molecular Weight : 241.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023518.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023518.html
Availability details for
Quantity
  • Availability
  • Add to Cart