MAT113930298 Matrix Scientific

1-Naphthyl 3-piperidinyl ether

  • MDL number MFCD08688439
  • Cas Number : 946681-27-2
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/023223.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023223.html
Availability details for
Quantity
  • Availability
  • Add to Cart