MAT113930399 Matrix Scientific

2-(Tetrahydro-2H-pyran-2-ylmethoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD08688548
  • Cas Number : 946727-96-4
  • Molecular Weight : 275.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022041.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022041.html
Availability details for
Quantity
  • Availability
  • Add to Cart