MAT113930403 Matrix Scientific

Tin trifluoromethanesulfonate

  • MDL number MFCD00191251
  • Cas Number : 62086-04-8
  • Molecular Weight : 416.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/006376.html

Safety Information

hcodes Toxic/Moisture Sensitive
MSDS http://www.matrixscientific.com/006376.html
Availability details for
Quantity
  • Availability
  • Add to Cart