MAT113930457 Matrix Scientific

3-Phenyl-2-(1H-pyrrol-1-yl)propanoic acid

  • MDL number MFCD03856011
  • Cas Number : 46687-57-4
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024187.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024187.html
Availability details for
Quantity
  • Availability
  • Add to Cart