MAT113930492 Matrix Scientific

2-Piperidin-1-yl-propylamine

  • MDL number MFCD06446934
  • Cas Number : 54151-70-1
  • Molecular Weight : 142.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021514.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021514.html
Availability details for
Quantity
  • Availability
  • Add to Cart