MAT113930580 Matrix Scientific

2-(2-Toluidino)acetohydrazide

  • MDL number MFCD01536452
  • Cas Number : 112255-66-0
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023960.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023960.html
Availability details for
Quantity
  • Availability
  • Add to Cart