MAT113930581 Matrix Scientific

3-m-Tolyloxy-propylamine

  • MDL number MFCD08271804
  • Cas Number : 26646-51-5
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021624.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021624.html
Availability details for
Quantity
  • Availability
  • Add to Cart