MAT113930592 Matrix Scientific

2-Trifluoromethyl-3H-benzoimidazole-5-carboxylic acid

  • MDL number MFCD01554219
  • Cas Number : 82791-93-3
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021589.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021589.html
Availability details for
Quantity
  • Availability
  • Add to Cart