MAT117644182 Matrix Scientific

N-[2-Fluoro-4-(methylsulfonyl)phenyl]-piperidin-4-amine hydrochloride

  • MDL number MFCD09064903
  • Cas Number : 1185128-34-0
  • Molecular Weight : 308.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016100.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016100.html
Availability details for
Quantity
  • Availability
  • Add to Cart