MAT117644195 Matrix Scientific

[2-(1H-Pyrazol-1-yl)propyl]aminedihydrochloride

  • MDL number MFCD06801245
  • Cas Number : 1184979-75-6
  • Molecular Weight : 198.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016538.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016538.html
Availability details for
Quantity
  • Availability
  • Add to Cart