MAT117644199 Matrix Scientific

1-Cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD04309657
  • Cas Number : 640264-45-5
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012120.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012120.html
Availability details for
Quantity
  • Availability
  • Add to Cart