MAT117644210 Matrix Scientific

5,6-Dimethyl-thieno[2,3-d]thiazol-2-ylaminehydrochloride

  • MDL number MFCD08447196
  • Cas Number : 1185303-27-8
  • Molecular Weight : 220.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/011386.html

Safety Information

MSDS http://www.matrixscientific.com/011386.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart