MAT117644217 Matrix Scientific

Ethyl [3,5-bis(trifluoromethyl)phenyl acetate]

  • MDL number MFCD08689894
  • Cas Number : 144632-97-3
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/018287.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018287.html
Availability details for
Quantity
  • Availability
  • Add to Cart