MAT117644219 Matrix Scientific

[(1-Ethylpiperidin-3-yl)methyl]amine

  • MDL number MFCD07757606
  • Cas Number : 102459-02-9
  • Molecular Weight : 142.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 89
Density 0.904/20
Country of Origin USA
Boiling point 86-87� Torr
Product Link http://www.matrixscientific.com/016359.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016359.html
Availability details for
Quantity
  • Availability
  • Add to Cart