MAT117644221 Matrix Scientific

1-(2-Fluoro-benzyl)-piperazinehydrochloride

  • MDL number MFCD05656276
  • Cas Number : 435345-41-8
  • Molecular Weight : 230.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008557.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008557.html
Availability details for
Quantity
  • Availability
  • Add to Cart