MAT117644235 Matrix Scientific

4-Fluoro-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD00061284
  • Cas Number : 67515-59-7
  • Molecular Weight : 189.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-69
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002276.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/002276.html
Availability details for
Quantity
  • Availability
  • Add to Cart