MAT117644236 Matrix Scientific

Furan-2-carboxylic acid (4-amino-2,5-dimethoxy-phenyl)-amide

  • MDL number MFCD02596288
  • Cas Number : 462067-17-0
  • Molecular Weight : 262.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008334.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008334.html
Availability details for
Quantity
  • Availability
  • Add to Cart