MAT117644242 Matrix Scientific

(1H-Indol-3-ylmethyl)-(2-methoxy-benzyl)-amine

  • MDL number MFCD01135668
  • Cas Number : 625409-25-8
  • Molecular Weight : 266.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009979.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009979.html
Availability details for
Quantity
  • Availability
  • Add to Cart