MAT117644246 Matrix Scientific

3-(2-Amino-thiazol-4-yl)-propionic acidmethyl ester hydrochloride

  • MDL number MFCD02017544
  • Cas Number : 436099-75-1
  • Molecular Weight : 222.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010270.html

Safety Information

MSDS http://www.matrixscientific.com/010270.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart