MAT117644271 Matrix Scientific

1-(2-Piperidin-1-yl-ethyl)-1H-benzoimidazol-2-yl-amine dihydrochloride

  • MDL number MFCD01578114
  • Cas Number : 435342-20-4
  • Molecular Weight : 317.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009959.html

Safety Information

MSDS http://www.matrixscientific.com/009959.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart