MAT117644280 Matrix Scientific

2-(2-Thiophen-2-yl-acetylamino)-benzoic acid

  • MDL number MFCD00980015
  • Cas Number : 330635-60-4
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/009908.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/009908.html
Availability details for
Quantity
  • Availability
  • Add to Cart