MAT117644285 Matrix Scientific

6-(Trifluoromethyl)pyridine-3-amidoxime

  • MDL number MFCD06245538
  • Cas Number : 386704-16-1
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-195
Country of Origin USA
Product Link http://www.matrixscientific.com/006825.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006825.html
Availability details for
Quantity
  • Availability
  • Add to Cart