MAT117958527 Matrix Scientific

2-Aza-spiro[5.5]undecanehydrochloride

  • MDL number MFCD00078172
  • Cas Number : 1185298-18-3
  • Molecular Weight : 189.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010336.html

Safety Information

MSDS http://www.matrixscientific.com/010336.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart