MAT117958529 Matrix Scientific

3-[(2-Cyclohexylethoxy)methyl]piperidinehydrochloride

  • MDL number MFCD09475404
  • Cas Number : 1185304-10-2
  • Molecular Weight : 261.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014464.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014464.html
Availability details for
Quantity
  • Availability
  • Add to Cart