MAT117958530 Matrix Scientific

3-[(Phenethyloxy)methyl]piperidinehydrochloride

  • MDL number MFCD09475405
  • Cas Number : 1093653-12-3
  • Molecular Weight : 255.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014489.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014489.html
Availability details for
Quantity
  • Availability
  • Add to Cart