MAT117958536 Matrix Scientific

4-(4-Chlorophenoxy)-3-(trifluoromethyl)anilinehydrochloride

  • MDL number MFCD09475412
  • Cas Number : 1185143-03-6
  • Molecular Weight : 324.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022535.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022535.html
Availability details for
Quantity
  • Availability
  • Add to Cart