MAT117958541 Matrix Scientific

4-(2-Propynyloxy)piperidinehydrochloride

  • MDL number MFCD09475417
  • Cas Number : 1185100-09-7
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023248.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023248.html
Availability details for
Quantity
  • Availability
  • Add to Cart