MAT117958553 Matrix Scientific

4-(Propoxymethyl)piperidine hydrochloride

  • MDL number MFCD09444901
  • Cas Number : 1051919-40-4
  • Molecular Weight : 193.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014591.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014591.html
Availability details for
Quantity
  • Availability
  • Add to Cart