MAT117958555 Matrix Scientific

3-[(3-Phenylpropoxy)methyl]piperidinehydrochloride

  • MDL number MFCD08447192
  • Cas Number : 1050509-62-0
  • Molecular Weight : 269.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014689.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014689.html
Availability details for
Quantity
  • Availability
  • Add to Cart