MAT117958576 Matrix Scientific

2-(1H-Indol-3-ylsulfanyl)-propionic acid

  • MDL number MFCD00981506
  • Cas Number : 63321-71-1
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/020191.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020191.html
Availability details for
Quantity
  • Availability
  • Add to Cart