MAT117958597 Matrix Scientific

4-(3-Methoxy-3-methyl-butoxy)-phenylamine

  • MDL number MFCD08144820
  • Cas Number : 883545-65-1
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020215.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020215.html
Availability details for
Quantity
  • Availability
  • Add to Cart