MAT117958606 Matrix Scientific

Methyl-[2-(tetrahydro-pyran-2-ylmethoxy)-ethyl]-amine

  • MDL number MFCD08146659
  • Cas Number : 883544-87-4
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020224.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020224.html
Availability details for
Quantity
  • Availability
  • Add to Cart