MAT117958615 Matrix Scientific

N'-Benzo[1,3]dioxol-5-ylmethyl-N,N-dimethyl-ethane-1,2-diamine

  • MDL number MFCD07364930
  • Cas Number : 500566-88-1
  • Molecular Weight : 222.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020233.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020233.html
Availability details for
Quantity
  • Availability
  • Add to Cart