MAT117958616 Matrix Scientific

Benzo[1,3]dioxol-5-ylmethyl-(2-methoxy-ethyl)-amine

  • MDL number MFCD07364933
  • Cas Number : 117986-59-1
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020234.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020234.html
Availability details for
Quantity
  • Availability
  • Add to Cart