MAT117958645 Matrix Scientific

(2,5,6-Trimethyl-4-oxo-4H-thieno[2,3-d]pyrimidin-3-yl)-acetic acid

  • MDL number MFCD03030493
  • Cas Number : 878657-12-6
  • Molecular Weight : 252.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020273.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020273.html
Availability details for
Quantity
  • Availability
  • Add to Cart