MAT117958663 Matrix Scientific

2-(4-Chloro-3,5-dimethyl-pyrazol-1-yl)-ethylamine

  • MDL number MFCD04967723
  • Cas Number : 925634-46-4
  • Molecular Weight : 173.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020294.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020294.html
Availability details for
Quantity
  • Availability
  • Add to Cart