MAT117958667 Matrix Scientific

(2-Pyridin-4-yl-thiazol-4-yl)-acetic acid

  • MDL number MFCD06739822
  • Cas Number : 31112-92-2
  • Molecular Weight : 220.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020298.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020298.html
Availability details for
Quantity
  • Availability
  • Add to Cart