MAT117958680 Matrix Scientific

1-(2-Methoxy-benzyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD07801162
  • Cas Number : 876892-41-0
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020313.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020313.html
Availability details for
Quantity
  • Availability
  • Add to Cart