MAT128014516 Matrix Scientific

5-Cyano-2-fluoropyridine

  • MDL number MFCD09261087
  • Cas Number : 3939-12-6
  • Molecular Weight : 122.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/024640.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/024640.html
Availability details for
Quantity
  • Availability
  • Add to Cart