MAT128014562 Matrix Scientific

2-(1H-Pyrazol-4-yl)-ethylamine dihydrochloride

  • MDL number MFCD09800617
  • Cas Number : 6429-11-4
  • Molecular Weight : 184.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-228
Country of Origin USA
Product Link http://www.matrixscientific.com/019667.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019667.html
Availability details for
Quantity
  • Availability
  • Add to Cart