MAT152385955 Matrix Scientific

4-(2-Benzylphenoxy)aniline hydrochloride

  • MDL number MFCD09879180
  • Cas Number : 1185297-18-0
  • Molecular Weight : 311.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022444.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022444.html
Availability details for
Quantity
  • Availability
  • Add to Cart