MAT152385965 Matrix Scientific

3-[(3-Chlorophenoxy)methyl]piperidinehydrochloride

  • MDL number MFCD09879191
  • Cas Number : 1185297-64-6
  • Molecular Weight : 262.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023001.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023001.html
Availability details for
Quantity
  • Availability
  • Add to Cart