MAT152385973 Matrix Scientific

Methyl 2-[4-(4-piperidinylmethoxy)phenyl]acetatehydrochloride

  • MDL number MFCD09879204
  • Cas Number : 1185153-80-3
  • Molecular Weight : 299.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023117.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023117.html
Availability details for
Quantity
  • Availability
  • Add to Cart