MAT152385978 Matrix Scientific

3-(3-Fluorophenoxy)piperidine hydrochloride

  • MDL number MFCD09879210
  • Cas Number : 1184976-95-1
  • Molecular Weight : 231.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023189.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023189.html
Availability details for
Quantity
  • Availability
  • Add to Cart