MAT152386004 Matrix Scientific

2-Naphthyl 3-pyrrolidinyl ether hydrochloride

  • MDL number MFCD09879247
  • Cas Number : 127342-10-3
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/023421.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023421.html
Availability details for
Quantity
  • Availability
  • Add to Cart